4-Hydroxy-2-(methylsulphanyl)-6-phenyl-5-pyrimidinecarbonitrile
Catalog # APOSOR111606-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.284 g/mol
Storage Temperature:Ambient
CAS Number:97693-21-5
Formula:C₁₂H₉N₃OS
MDL Number:MFCD00974848