Order Entry
United Kingdom
ContactUsLinkComponent
 
Corrosive
4-(Chloromethyl)-7-methoxy-2H-chromen-2-one
Catalog # APOSOR111001-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
4-(Chloromethyl)-7-methoxy-2H-chromen-2-one
Catalog # APOSOR111001-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :224.64 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
CAS Number:41295-55-0
Formula:C₁₁H₉ClO₃
MDL Number:MFCD03731259