Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4'-Methyl-3-chloropropiophenone
4'-Methyl-3-chloropropiophenone
4'-Methyl-3-chloropropiophenone
APOSOR11066-1G
:
Apollo Scientific
:
4'-Methyl-3-chloropropiophenone
APOSOR11066-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
182.647 g/mol
:
GHS07
:
Ambient
:
22422-21-5
:
C₁₀H₁₁ClO
:
MFCD00028195