Order Entry
United Kingdom
ContactUsLinkComponent
2-Ethyl[1,3]oxazolo[4,5-b]pyridine 95%
2-Ethyl[1,3]oxazolo[4,5-b]pyridine 95%
Catalog # APOSOR110179-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Ethyl[1,3]oxazolo[4,5-b]pyridine 95%
Catalog # APOSOR110179-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :148.162 g/mol
Storage Temperature:Ambient
CAS Number:52333-88-7
Formula:C₈H₈N₂O
MDL Number:MFCD11110208