
(+)-Bis(pinanediolato)diboron
Catalog # APOSOR10609-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:358.088 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:230299-05-5
Formula:C₂₀H₃₂B₂O₄
MDL Number:MFCD27664803