2,3,4,6-Tetra-O-acetyl-2-hydroxy-D-glucal ≥98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1,5-Anhydro-D-arabino-hex-1-enitol 2,3,4,6-tetraacetate
CAS Number:3366-47-0
:
Formula:C₁₄H₁₈O₉
MDL Number:MFCD00040568
Molecular Weight:330.287 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together