2-Azidoethyl 2,3,4,6-tetra-O-acetyl-α-D-mannopyranoside ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:140428-83-7
:
Formula:C₁₆H₂₃N₃O₁₀
MDL Number:MFCD19981031
Molecular Weight:417.368 g/mol
Melting Pt:80…82 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together