Maleimidopropionic acid hydrazide hydrochloride
Catalog # APOSBICL230-50MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:219.626 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:293298-33-6
Formula:C₇H₉N₃O₃·ClH
MDL Number:MFCD00467760