

5-Bromo-4-chloro-(1H-indol-3-yl) butanoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:129541-43-1
:
Formula:C₁₂H₁₁BrClNO₂
MDL Number:MFCD00083261
:316.578 g/mol
:GHS05
:GHS07
Storage Temperature:Freezer
Frequently Bought Together