Polyethylene glycol 4,000, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:25322-68-3
:MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779612
:4000 g/mol
Storage Temperature:Ambient
Specification Test Results
- Average molecular weight:3500 - 4500
- Hydroxyl value:25 - 32
- pH (5%):4,0 - 8,0
- Viscosity (20°C, 50% water):110 - 142 mPa.s
- Melting point (clear):about 55°C
- Identification (IR):passes test
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC26609.290 spec_VWRC26609.290 spec_VWRC26609.460
Product specification
Frequently Bought Together