GSK-583 95%
Catalog # ACRO468211000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to yellow Powder or solid
- HPLC:≥94%
- MS:complies
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:398.46 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:1346547-00-9
Formula:C₂₀H₁₉FN₄O₂S
MDL Number:MFCD30343846