
I-BET151 95%
Catalog # ACRO467841000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow to light brown Powder or solid
- HPLC:≥94%
- MS:complies
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:415.4445 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:1300031-49-5
Formula:C₂₃H₂₁N₅O₃
MDL Number:MFCD22124472