Nile blue A, certified by the Biological Stain Commission, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
Synonyms: Nile blue A sulphate, Nile blue sulphate, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium sulphate, Basic Blue 12
CAS Number:3625-57-8
:
Formula:C₄₀H₄₀N₆O₆S
MDL Number:MFCD00064529
Molecular Weight:732.84 g/mol
Melting Pt:>300 °C (dec., lit.)
Storage Temperature:Room Temperature
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Form):Powder
- Infrared Spectrum:Conforms to Structure
- Solubility (Colour)
1 mg/mL, H2O:Dark Blue - Dye Content:≥75 %
- Biological Stain Commission:Commission Certified Number: AcNb-21
Specification Test Results
Frequently Bought Together