Biebrich Scarlet : Acid Fuchsin solution, Sigma-Aldrich®
Catalog # HT151-250ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:250 mL
- Seq:2590
- Environmentally Preferable:
Specifications
About this item
:
Storage Temperature:Ambient