Order Entry
United Kingdom
ContactUsLinkComponent
Indocyanine green, Sigma-Aldrich®
Irritant
Indocyanine green, Sigma-Aldrich®
Certificates
undefined
Irritant
Indocyanine green, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    Green to very dark green and brown to very dark brown
  • Appearance (Form):
    Powder
  • Solubility (Colour):
    0.01 g in 10 ml water
  • Solubility (Method):
    0.01 g in 10 ml water
  • Nitrogen Content:
    2.7 - 3.6 %
  • Dye Content:
    ≥75 %
  • Recommended Retest Period:
    72 Months

Specification Test Results

Frequently Bought Together