Proflavine hemisulphate hydrate, powder, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:1811-28-5
:MFCD00013150
Formula:(C₁₃H₁₁N₃)₂·H₂SO₄·xH₂O
MDL Number:MFCD00149650
:516.58 g/mol (anhy)
:M-Clarity™ quality level = MQ100
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):Faint Orange to Red-Brown to Brown
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Water (by Karl Fischer):≤10.0 %
- Solubility (Colour):Faint Orange to Dark Brown
- Solubility (Form)
10 mg/ml, H2O:Solution - Carbon
Anhydrous:59.2 - 61.7 % - Nitrogen
Anhydrous:15.9 - 16.6 %
Specification Test Results
Frequently Bought Together