Order Entry
United Kingdom
ContactUsLinkComponent
Surface disinfectants, liquid, Spor-Klenz®
Surface disinfectants, liquid, Spor-Klenz®
Catalog # STER6525M2EC
Supplier:  STERIS
Surface disinfectants, liquid, Spor-Klenz®
Catalog # STER6525M2EC
Supplier:  STERIS
Specifications
  • Description:
    Ready-to-use cold sterilant
  • Seq:
    01
  • Environmentally Preferable:
  • Pk Info:
    4 x 850 mL
  • Pk:
    4
Specifications
About this item
Spor-Klenz® Ready-To-Use Sterilants are fast acting, liquid sporicides specifically formulated for sterilization and disinfection of environmental, hard non-porous surfaces. Sterilants contain the proven blend of peracetic acid, hydrogen peroxide and acetic acid, that provides fast, effective microbial control of challenging spores such as Bacillus subtilis and Bacillus cereus and fungi like Aspergillus brasiliensis.
  • Formulation is ready to use without any mixing or activation
  • Excellent material compatibility, allows use on most surfaces
  • Supports worker safety
  • Low toxicity profile
  • Eliminates errors in dilution preparation and the need to validate preparation procedures
  • Manufactured in accordance with cGMP with complete lot traceability
This formulation offers operational flexibility by being diluted with water for use as a disinfectant or sanitizer.
Caution: Use biocides safely. Always read the label and product information before use.