Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Azelastine Hydrochloride

Azelastine Hydrochloride

Irritant
Azelastine Hydrochloride
Certificates
Azelastine Hydrochloride

2 Options Available Below

Product Details & Documents
  • Bioactive
Synonyms:  4-(4-Chlorobenzyl)-2-(1-methylazepan-4-yl)phthalazin-1(2H)-one hydrochloride, Azelastine HCl, 4-(4-Chlorobenzyl)-2-(1-methylazepan-4-yl)-1-phthalazinone Hydrochloride
  • CAS Number:79307-93-0
  • Formula:C₂₂H₂₄ClN₃O
  • MDL Number:MFCD00242783
  • Molecular Weight:418.36 g/mol
  • Melting Pt:225…229 °C
  • Storage Temperature:Freezer
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    224 to 228 °C
  • HPLC:
    ≥97.5 %
  • Titration Argentimetric:
    97.5 to 102.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - £46.90
    £46.90
    1 * 250 mg
       £46.90

  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - £139.00
    £139.00
    1 * 1 g
       £139.00