Order Entry
United Kingdom
ContactUsLinkComponent
2,2,4,6,6-Pentamethylheptane
Flammable
Health
2,2,4,6,6-Pentamethylheptane
Certificates
undefined
Flammable
Health
2,2,4,6,6-Pentamethylheptane

Specification Test Results

  • Appearance:
    Clear almost colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4170 to 1.4200 (20°C, 589 nm)
  • GC:
    ≥94 %

Specification Test Results

Frequently Bought Together