genesig® SARS-COV-2 winterplex real-time PCR assay
Catalog # D00020
Supplier: PRIMERDESIGN
CAS Number:
Specifications
- Pk:96 Tests
- Description:genesig® SARS-COV-2 Winterplex for Real-Time PCR Assay
Specifications
About this item
Multiplex qPCR assay for discrimination and qualitative detection of RNA from SARS-CoV-2 (ORF1ab and S gene targets), Influenza A, Influenza B and RSV A&B. With discrimination between these 4 winter pathogens in conjunction with an extremely high level of clinical sensitivity, this IVD provides the ideal flexible solution for total control, specifically designed for winter 2020/21.
- Highly sensitive for all targets (each target ≤ 1 copy/ul)
- 96 tests
- Validated against CFX (Bio-Rad), ABI 7500 (Applied Biosystems) and LC480 II (Roche)
- Master mix included
- Traceability of values assigned to calibrating of control materials. Assured through reference measurement procedures
- Comprehensive validation in concordance with ISO13485. Post-market maintenance of technical files, including strain information
- Total kit performance measurement and assurance provided
- Open platform chemistry, with calibration panel available for instruments requiring colour compensation (Roche LC480 II)
Certifications: CE-IVD
Delivery: Lyophilised kits with no dry ice shipping requirement.