Order Entry
United Kingdom
ContactUsLinkComponent
2,6-Dichloroquinone-4-chloroimide ≥98%, AnalaR NORMAPUR®
Flammable
Irritant
2,6-Dichloroquinone-4-chloroimide ≥98%, AnalaR NORMAPUR®
Catalog # 88253.120
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
2,6-Dichloroquinone-4-chloroimide ≥98%, AnalaR NORMAPUR®
Catalog # 88253.120
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Assay:
    ≥ 98 %
  • Appearance: Brownish-yellow crystalline powder :
    Passes test
  • Melting point :
    65 - 68 °C
  • Spec.absorptivity (312 nm; 0.001%; ethanol) :
    ≥ 800

Specification Test Results

About this item

 :
UN:3224
ADR:4.1,
 :GHS02
 :GHS07
CAS Number:101-38-2
Formula:C₆H₂Cl₃NO
MDL Number:MFCD00001611