Centrifuge, refrigerated, floor-standing, Sorvall™ RC-6 Plus superspeed
Supplier: Thermo Fisher Scientific
Certificates
About this item
The Sorvall™ RC-6 Plus superspeed centrifuge combines superior functionality, unparalleled safety and traditional Sorvall™ reliability in a compact, user-friendly design.
- Non bolt-down design allowing the centrifuge to be placed within the lab without being anchored it to the floor
- Intuitive controls and dual display
- 30-program memory allowing common protocols to be stored and retrieved easily, and offering flexibility in a multi-user environment
- GMP/GLP-compatible data management (Traceability and quality control for Biotech/Pharmaceutical processing needs)
Offering 4-liter capacity and speeds of up to 22000 min⁻¹ (55200 ×g), the Sorvall™ RC-6 Plus is a high-productivity solution that delivers maximum throughput and minimum maintenance.
Ordering information: Other rotors available on request.