£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
Standards
Organic Standards
Analytical standard, Lufenuron, reference material, Fluka™
Analytical standard, Lufenuron, reference material, Fluka™
HONCR1732-100MG
:
Honeywell Chemicals
:
Analytical standard, Lufenuron, reference material, Fluka™
HONCR1732-100MG
:
Honeywell Chemicals
:
Pack type:
Glass bottle
Pk:
100 mg
Standards:
Reference material
CAS No.:
103055-07-8
Description:
Lufenuron
Description:
Lufenuron
Form:
Neat