

Chromium(III) acetylacetonate for synthesis, Sigma-Aldrich®
Catalog # 8.02485.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:11 g/l (20 °C)
- Melting Point:213 - 216 °C
- Molar Mass:349.33 g/mol
- Bulk Density:450 kg/m3
- Boiling Point:339 - 341 °C
- pH:6 (1 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:349.32 g/mol
Boiling Pt:340 °C (1013 hPa)
Melting Pt:215.85 °C
Density:1.266 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:244-526-0
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:21679-31-2
Formula:C₁₅H₂₁CrO₆
MDL Number:MFCD00000015