Order Entry
United Kingdom
ContactUsLinkComponent
Chromium(III) acetylacetonate 97%
Irritant
Environment
Chromium(III) acetylacetonate 97%
Certificates
undefined
Irritant
Environment
Chromium(III) acetylacetonate 97%

Specification Test Results

  • Appearance:
    Brown to purple Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Assay:
    14.4 to 15.3 % (Cr)

Specification Test Results

Related Information

Frequently Bought Together