
Dansyl chloride, Sigma-Aldrich®
Catalog # 311155-1
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:269.75 g/mol
Merck Index:13,02840
Melting Pt:69…73 °C
Storage Temperature:Refrigerator
EINECS:210-092-6
UN:3261
ADR:8,III
:GHS05
CAS Number:605-65-2
Formula:C₁₂H₁₂ClNO₂S
MDL Number:MFCD00003985