Methyl-1-hydroxy-2-naphthoate, Sigma-Aldrich®
Catalog # SIAL540188-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:202.21 g/mol
:M-Clarity™ quality level = MQ100
EINECS:213-434-2
CAS Number:948-03-8
Formula:C₁₂H₁₀O₃
MDL Number:MFCD00043823