

Tetraethylammonium cyanide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13435-20-6
:
Formula:C₉H₂₀N₂
:156.27 g/mol
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:236-566-2
UN:2811
:GHS06
:GHS09
Storage Temperature:Ambient
Frequently Bought Together