Sand (sea sand) for analysis, purified by acid, calcinated, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:14808-60-7
:7631-86-9
Formula:SiO₂
Boiling Pt:>2200 °C (1013 hPa)
Density:2.65 g/cm³ (20 °C)
Molecular Weight:60.08 g/mol
Melting Pt:1713 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-545-4
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Substances soluble in hydrochloric acid:≤ 0.2 %
- Chloride (Cl):≤ 0.005 %
- Acid-soluble Iron (Fe):≤ 0.01 %
- Loss on ignition (900 °C):≤ 0.05 %
- Particle size (0.1 - 0.315 mm):≥ 90 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.07712.5000_TDS_EN ProductPage_107712
Product specification
Merck Quality Materials for Absorption, Adsorption and Filtration
Product Literature
msds_107712
SDS
Frequently Bought Together