Indium(III) acetylacetonate, Sigma-Aldrich®
Catalog # SIALI3300-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:412.15 g/mol
:M-Clarity™ quality level = MQ100
EINECS:238-378-6
CAS Number:14405-45-9
Formula:C₁₅H₂₁InO₆
MDL Number:MFCD00013494