Order Entry
United Kingdom
ContactUsLinkComponent
 
Potassium hydrogen carbonate ≥99.95% (trace metals basis) (99.7-100.5% dry basis)
Catalog # 431583-250G
CAS Number:  
Potassium hydrogen carbonate ≥99.95% (trace metals basis) (99.7-100.5% dry basis)
Catalog # 431583-250G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (excluding dried substances):
    99.7 - 100.5 %
  • Water insoluble matter:
    Max. 0.01 %
  • Calcium (Ca):
    Max. 0.002 %
  • Iron (Fe):
    Max. 5.0 ppm
  • Magnesium (Mg):
    Max. 0.001 %
  • Sodium (Na):
    Max. 0.03 %
  • Metal trace analysis (ICP):
    Max. 500.0 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • S-compounds (as SO₄):
    Max. 0.003 %
  • Ammonium (NH₄):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.001 %
  • Phosphate (PO₄):
    Max. 5 ppm

Specification Test Results

About this item

 :
 :100.12 g/mol
Merck Index:13,07691
Melting Pt:100-120 °C (dec.)
Melting Pt:100…120 °C (dec.)
Density:2.17 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-059-0
CAS Number:298-14-6
Formula:KHCO₃
MDL Number:MFCD00011402