
Potassium carbonate sesquihydrate, Suprapur® for trace analysis, Supelco®
1.04926.0050
: Merck
:
- Pack type:Plastic bottle
- Pk:50 g
- Seq:3368
- Solubility:1300 g/l (20 °C)
- Melting Point:891 °C (anhydrous substance)
- Molar Mass:138.21 g/mol (anhydrous)
- pH:(50 g/l, H2O, 20 °C) strongly alkaline
:
:165.23 g/mol
:891 °C
:2.29 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:209-529-3
:GHS07
:Ambient
:6381-79-9
:CK₂O₃·1,5H₂O
:MFCD00149925