Bismuth sodium trioxide ≥80%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12232-99-4
:
Formula:BiNaO₃
MDL Number:MFCD00003474
Merck Index:14,8585
:279.97 g/mol
EINECS:235-455-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together