
meta-Phosphoric acid 33.5-36.5%, chips ACS, Fluka™
Catalog # 239275-2.5KG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg
- Seq:4076
Specifications
Specification Test Results
- Form:chips
- Identity (IR):complying
- Assay of (HPO₃)n:33.5 - 36.5 %
- Assay of (naPO₃)n:57.0 - 63.0 %
- Arsenic (As):max. 1 ppm
- Iron (Fe):max. 0.005 %
- Heavy metals (as Pb):max. 0.005 %
- Chloride (Cl):max. 0.001 %
- Nitrate (NO₃):max. 0.001 %
- Sulfate (SO₄):max. 0.005 %
- KMnO₄ reducing matter (as H₃PO₃):max. 0.02 %
Specification Test Results
About this item
:
:79.98 g/mol
Boiling Pt:600 °C(1,013 hPa)
Melting Pt:200 °C
Density:2.0 g/cm³
Storage Temperature:Room Temperature
EINECS:253-433-4
UN:3260
ADR:8,II
:GHS05
CAS Number:37267-86-0
Formula:(HPO₃)n
MDL Number:MFCD00011351