Cyclohexanone oxime, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
- CAS Number:100-64-1
- Formula:C₆H₁₁NO
- MDL Number:MFCD00001660
- Molecular Weight:113.16 g/mol
- Boiling Pt:206 °C (1013 hPa)
- Melting Pt:88…90 °C
- Density:1.13 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- EINECS:202-874-0
2 Options Available Below
Product Details & Documents
- CAS Number:100-64-1
- Formula:C₆H₁₁NO
- MDL Number:MFCD00001660
- Molecular Weight:113.16 g/mol
- Boiling Pt:206 °C (1013 hPa)
- Melting Pt:88…90 °C
- Density:1.13 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- EINECS:202-874-0



