Order Entry
United Kingdom
ContactUsLinkComponent
4'-Isopropylacetophenone for synthesis, Sigma-Aldrich®
4'-Isopropylacetophenone for synthesis, Sigma-Aldrich®
Catalog # 8.41367.0010
Supplier:  Merck
CAS Number:  
4'-Isopropylacetophenone for synthesis, Sigma-Aldrich®
Catalog # 8.41367.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 ml
  • Seq:
    2275

Specifications

Specification Test Results

  • Molar Mass:
    162.23 g/mol
  • Boiling Point:
    119 - 120 °C (13.3 hPa)
  • Density:
    0.970 g/cm3 (20 °C)
  • Hill Formula:
    C11H14O

Specification Test Results

About this item

 :
Density:0.97 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :162.23
EINECS:211-433-1
CAS Number:645-13-6
Formula:C₁₁H₁₄O
MDL Number:MFCD00048297