4'-Isopropylacetophenone
1 g
Catalog # FLUO022395-1G
Marketsource
Supplier: FLUOROCHEM
4'-Isopropylacetophenone
Catalog # FLUO022395-1G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 645-13-6
Jump to:
Product Details & Documents
Synonyms: p-Isopropylacetophenone, 4-Isopropylacetophenone
- CAS Number:645-13-6
- Formula:C₁₁H₁₄O
- MDL Number:MFCD00048297
- Molecular Weight:162.23
- Density:0.97 g/cm³ (20 °C)
- :Density: 0.97 g/cm³ (20 °C)
- EINECS:211-433-1
Specifications
Specifications
- Pk:1 g



