Eusolex® 4360
Catalog # 1.05376.0101
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
:
EINECS:205-031-5
UN:3077
:GHS09
Storage Temperature:Ambient
CAS Number:131-57-7
Formula:C₁₄H₁₂O₃
Density:1.3397 g/cm³ (25 °C)
ADR:9 III