Order Entry
United Kingdom
ContactUsLinkComponent
 
2-Methyl-3-nitrophenyl isocyanate ≥98%
Catalog # ABCRAB179055.0005
Supplier:  abcr
CAS Number:  
2-Methyl-3-nitrophenyl isocyanate ≥98%
Catalog # ABCRAB179055.0005
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :178.15 g/mol
Boiling Pt:118 °C (2.8 mmHg)
Melting Pt:34…37 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
CAS Number:23695-15-0
Formula:C₈H₆N₂O₃
MDL Number:MFCD00037046