3'-Fluoro-4'-hydroxyacetophenone
Marketsource
Supplier: FLUOROCHEM
Jump to:
About this item
Synonyms: 1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone
- CAS Number:403-14-5
- Formula:C₈H₇FO₂
- MDL Number:MFCD00215840
- Molecular Weight:154.14 g/mol
2 Options Available Below
Product Details & Documents
Synonyms: 1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone
- CAS Number:403-14-5
- Formula:C₈H₇FO₂
- MDL Number:MFCD00215840
- Molecular Weight:154.14 g/mol
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



