Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
ω-Bromo-3-nitroacetophenone

ω-Bromo-3-nitroacetophenone

Glass bottle for solids, 25 g
Catalog # MOLE15592918-25G
Marketsource
Supplier:   Molekula
ω-Bromo-3-nitroacetophenone
Catalog # MOLE15592918-25G
Supplier:  Molekula
Marketsource
CAS Number:  2227-64-7
ω-Bromo-3-nitroacetophenone

Glass bottle for solids, 25 g

CAS Number:  2227-64-7
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-Nitrophenacyl bromide, 2-Bromo-3'-nitroacetophenone, α-Bromo-3-nitroacetophenone, Bromomethyl m-nitrophenyl ketone
  • CAS Number:2227-64-7
  • Formula:C₈H₆BrNO₃
  • MDL Number:MFCD00024512
  • Molecular Weight:244.04 g/mol
  • Melting Pt:90…94 °C
  • EINECS:218-764-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.