ω-Bromo-3-nitroacetophenone
Glass bottle for solids, 50 g
Catalog # FLUO011206-50G
Marketsource
Supplier: FLUOROCHEM
ω-Bromo-3-nitroacetophenone
Catalog # FLUO011206-50G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 2227-64-7
Jump to:
Product Details & Documents
Synonyms: 3-Nitrophenacyl bromide, 2-Bromo-3'-nitroacetophenone, α-Bromo-3-nitroacetophenone, Bromomethyl m-nitrophenyl ketone
- CAS Number:2227-64-7
- Formula:C₈H₆BrNO₃
- MDL Number:MFCD00024512
- Molecular Weight:244.04 g/mol
- Melting Pt:90…94 °C
- EINECS:218-764-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Cat. No.:FLUO011206-50G



