MES sodium salt
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Sodium 2-Morpholinoethanesulphonate, Sodium 3-morpholin-1-ylethylsulphonate, 2-(N-Morpholino)ethanesulphonic acid sodium salt, Sodium 2-morpholinoethane-1-sulphonate, MES-Na
- CAS Number:71119-23-8
- Formula:C₆H₁₂NNaO₄S
- MDL Number:MFCD00065473
- Molecular Weight:217.22 g/mol
- EINECS:275-203-2
2 Options Available Below
Product Details & Documents
Synonyms: Sodium 2-Morpholinoethanesulphonate, Sodium 3-morpholin-1-ylethylsulphonate, 2-(N-Morpholino)ethanesulphonic acid sodium salt, Sodium 2-morpholinoethane-1-sulphonate, MES-Na
- CAS Number:71119-23-8
- Formula:C₆H₁₂NNaO₄S
- MDL Number:MFCD00065473
- Molecular Weight:217.22 g/mol
- EINECS:275-203-2
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



