
Phenylacetaldehyde 98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:122-78-1
:
Formula:C₈H₈O
MDL Number:MFCD00006993
Merck Index:12,07419
Boiling Pt:195 °C (1013 hPa)
Melting Pt:33…34 °C
Density:1.029 g/cm³
Flash Pt:68 °C
:120.15 g/mol
EINECS:204-574-5
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5250 to 1.5350 (20°C, 589 nm)
- GC:≥97.5 %
- Stabilizer:Approx. 0.01 % Citric acid
- Relative density:(20/4°C) 1.026 to 1.036
- Acid value:≤5 mg KOH/g
Specification Test Results
Frequently Bought Together