Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sodium-2-ethylhexanoate ≥97%

Sodium-2-ethylhexanoate ≥97%

Health
Sodium-2-ethylhexanoate ≥97%
Certificates
Sodium-2-ethylhexanoate ≥97%
Sodium-2-ethylhexanoate ≥97%

3 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 19766-89-3 Molecular Formula: C8H15NaO2 Formula Weight: 166.20 Odour: Odourless Melting Point: 300° MDL No.: MFCD00014007

Synonyms:  2-Ethylcaproic acid sodium salt, Sodium 2-ethylhexanoate, 2-Ethylhexanoic acid sodium salt
  • CAS Number:19766-89-3
  • Formula:C₈H₁₅NaO₂
  • MDL Number:MFCD00014007
  • Molecular Weight:166.2 g/mol
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:243-283-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - £26.40
    £26.40
    1 * 250 g
       £26.40

  • Glass bottle for solids
    1.000 g
    Log in for availability
    Log in for availability
    1 * 1.000 g
    1 * 1.000 g - £77.40
    £77.40
    1 * 1.000 g
       £77.40

  • Glass bottle for solids
    5.000 g
    Log in for availability
    Log in for availability
    1 * 5.000 g
    1 * 5.000 g - £268.00
    £268.00
    1 * 5.000 g
       £268.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.