Diethyl butylmalonate, Sigma-Aldrich®
SIAL112038-100ML
: Merck
:
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
:
:216.28 g/mol
:13,01575
:235 °C (1013 hPa)
:0.976 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:205-089-1
:133-08-4
:C₁₁H₂₀O₄
:MFCD00009170