Order Entry
United Kingdom
ContactUsLinkComponent
 
D(-)-2-(2,5-Dihydrophenyl)glycine ≥97.0% (by titrimetric analysis)
Catalog # TCIAD2539-5G
Supplier:  TCI
CAS Number:  
D(-)-2-(2,5-Dihydrophenyl)glycine ≥97.0% (by titrimetric analysis)
Catalog # TCIAD2539-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Specific rotation [a]20/D:
    -150 to -160 deg (C=1, 1mol/L HCl)

Specification Test Results

About this item

Synonyms: (R)-2-Amino-2-(cyclohexa-1,4-dien-1-yl)acetic acid, (R)-(-)-2-Amino-2-(cyclohexa-1,4-dienyl)acetic Acid
 :
 :153.18 g/mol
EINECS:247-999-1
CAS Number:26774-88-9
Formula:C₈H₁₁NO₂
MDL Number:MFCD00137746