Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Sodium iodoacetate 99%

Sodium iodoacetate 99%

Corrosive
Toxic
Sodium iodoacetate 99%
Certificates
Sodium iodoacetate 99%

2 Options Available Below

Product Details & Documents
Synonyms:  Sodium 2-iodoacetate, Iodoacetic Acid, Sodium Salt
  • CAS Number:305-53-3
  • Formula:C₂H₂INaO₂
  • MDL Number:MFCD00002686
  • Molecular Weight:207.93 g/mol
  • Melting Pt:208…210 °C
  • Flash Pt:208 °C
  • Storage Temperature:Freezer
  • EINECS:206-165-7
  • UN:2811
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Light yellow to yellow-brown Powder
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥98.5 %
  • GC:
    ≥96.0 %
  • Stabilizer:
    hydroquinone: approx. 0.05 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - £93.10
    £93.10
    1 * 25 g
       £93.10

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £255.00
    £255.00
    1 * 100 g
       £255.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.