Order Entry
United Kingdom
ContactUsLinkComponent
2-Benzylimidazoline ≥97.0% (by GC, titration analysis)
2-Benzylimidazoline ≥97.0% (by GC, titration analysis)
Catalog # TCIAB3806-5G
Supplier:  TCI
CAS Number:  
2-Benzylimidazoline ≥97.0% (by GC, titration analysis)
Catalog # TCIAB3806-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to lump
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    64.0 to 68.0 deg-C

Specification Test Results

About this item

Synonyms: Tolazoline
 :
 :160.22 g/mol
EINECS:200-448-9
 :GHS07
Storage Temperature:Refrigerator
CAS Number:59-98-3
Formula:C₁₀H₁₂N₂
MDL Number:MFCD00005182
Boiling Pt:188°C
Merck Index:14,09506