Order Entry
United Kingdom
ContactUsLinkComponent
Methyl enantate
Methyl enantate
Catalog # EHERC14126900
Supplier:  EHRENSTORFER
CAS Number:  
Methyl enantate
Catalog # EHERC14126900
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
Merck Index:12,04695
Boiling Pt:172 °C (1013 hPa)
Melting Pt:–56 °C
Density:0.88 g/cm³ (20 °C)
 :144.21 g/mol
EINECS:203-428-8
CAS Number:106-73-0
Formula:C₈H₁₆O₂
MDL Number:MFCD00009537